α-Bromo-4-iodotoulene ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 4-Iodobenzyl bromide, 1-(Bromomethyl)-4-iodobenzene, α-Bromo-p-iodotoulene

I0571-25G I0571-5G
TCI0571-25GEA 278 USD
TCI0571-25G TCI0571-5G
α-Bromo-4-iodotoulene ≥97.0% (by GC)
α-Bromo-4-iodotoulene

CAS Number: 16004-15-2
MDL Number: MFCD00209656
Molecular Formula: C7H6BrI
Molecular Weight: 296.93
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 80
Storage Temperature: 0-10°C


Other Product Names / Synonyms: alpha-Bromo-4-iodotoluene

Formula: C₇H₆BrI
MW: 296.93 g/mol
Melting Pt: 80 °C
MDL Number: MFCD00209656
CAS Number: 16004-15-2
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acb9d7eedeb79626f61d20b3f1f78f02472d5bee&vid=acb9d7eedeb79626f61d20b3f1f78f02472d5bee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR