3,5-Bis(trifluoromethyl)phenyl isothiocyanate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
1,3-Bis(trifluoromethyl)-5-isothiocyanatobenzene, 1-Isothiocyanato-3,5-bis(trifluoromethyl)benzene
I0569-25G
I0569-5G
TCI0569-25GEA
195
USD
TCI0569-25G
TCI0569-5G
3,5-Bis(trifluoromethyl)phenyl isothiocyanate ≥98.0% (by GC)
3,5-Bis(trifluoromethyl)phenyl isothiocyanate
CAS Number: 23165-29-9
MDL Number: MFCD00040838
Molecular Formula: C9H3F6NS
Molecular Weight: 271.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 108
Specific Gravity (20/20): 1.48
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Isothiocyanic Acid 3,5-Bis(trifluoromethyl)phenyl Ester
Formula:
C₉H₃F₆NS MW: 271.19 g/mol Boiling Pt: 95 °C Density: 1.48 (20 °C) Flash Pt: 108 °C |
MDL Number:
MFCD00040838 CAS Number: 23165-29-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...