3-Chloro-4-methylphenyl isocyanate ≥97.0% (by GC)
Supplier: TCI America
CAS Number: 28479-22-3
MDL Number: MFCD00013858
Molecular Formula: C8H6ClNO
Molecular Weight: 167.59
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 124
Flash Point (°C): 109
Freezing point (°C): 23
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Isocyanic Acid 3-Chloro-4-methylphenyl Ester
Formula:
C₈H₆ClNO Boiling Pt: 124 °C Flash Pt: 109 °C |
MDL Number:
MFCD00013858 CAS Number: 28479-22-3 UN: 2236 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...