2,4-Dichlorophenyl isocyanate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,4-Dichlorophenylisocyantae
I0555-25G
I0555-5G
TCI0555-25GEA
145
USD
TCI0555-25G
TCI0555-5G
2,4-Dichlorophenyl isocyanate ≥98.0% (by GC, titration analysis)
2,4-Dichlorophenyl isocyanate
CAS Number: 2612-57-9
MDL Number: MFCD00002001
Molecular Formula: C7H3Cl2NO
Molecular Weight: 188.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 58
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Isocyanic Acid 2,4-Dichlorophenyl Ester
Formula:
C₇H₃Cl₂NO MW: 188.01 g/mol Melting Pt: 58 °C |
MDL Number:
MFCD00002001 CAS Number: 2612-57-9 UN: 2250 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...