3-Bromophenyl isocyanate ≥98.0% (by GC)

Supplier: TCI America

I0553-25G I0553-5G
TCI0553-25GEA 274.1 USD
TCI0553-25G TCI0553-5G
3-Bromophenyl isocyanate ≥98.0% (by GC)
3-Bromophenyl isocyanate

CAS Number: 23138-55-8
MDL Number: MFCD00002014
Molecular Formula: C7H4BrNO
Molecular Weight: 198.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 108
Specific Gravity (20/20): 1.59
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Isocyanic Acid 3-Bromophenyl Ester

Formula: C₇H₄BrNO
Boiling Pt: 220 °C
Density: 1.59 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00002014
CAS Number: 23138-55-8
UN: 2206
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83f2a1e258a68441fb925392d9109dc754b8ef7c&vid=83f2a1e258a68441fb925392d9109dc754b8ef7c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR