3-Methoxyphenyl isocyanate ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 1-Isocyanato-3-methoxybenzene

I0551-25G I0551-5G
TCI0551-25GEA 235 USD
TCI0551-25G TCI0551-5G
3-Methoxyphenyl isocyanate ≥97.0% (by GC)
3-Methoxyphenyl isocyanate

CAS Number: 18908-07-1
MDL Number: MFCD00002019
Molecular Formula: C8H7NO2
Molecular Weight: 149.15
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 96
Specific Gravity (20/20): 1.16
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Isocyanic Acid 3-Methoxyphenyl Ester

Formula: C₈H₇NO₂
MW: 149.15 g/mol
Boiling Pt: 95 °C
Density: 1.16 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00002019
CAS Number: 18908-07-1
UN: 2206
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61cf33b8754c3bbecb428eede4f6db5c2aa6000b&vid=61cf33b8754c3bbecb428eede4f6db5c2aa6000b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR