4-Iodobenzoyl chloride ≥98.0%

Supplier: TCI America

Synonyms: p-Iodobenzoyl chloride

I0508-25G I0508-5G
TCI0508-25GEA 155 USD
TCI0508-25G TCI0508-5G
4-Iodobenzoyl chloride ≥98.0%
4-Iodobenzoyl chloride

CAS Number: 1711-02-0
MDL Number: MFCD00001058
Molecular Formula: C7H4ClIO
Molecular Weight: 266.46
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 100
Melting point (°C): 65
Flash Point (°C): 110

Formula: C₇H₄ClIO
MW: 266.47 g/mol
Boiling Pt: 100
Melting Pt: 65 °C
Flash Pt: 110 °C
MDL Number: MFCD00001058
CAS Number: 1711-02-0
UN: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7618a56308f9bedde138b687d1c55e5d49867192&vid=7618a56308f9bedde138b687d1c55e5d49867192&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR