4-Methoxyphenyl isocyanate ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 5416-93-3
MDL Number: MFCD00002026
Molecular Formula: C8H7NO2
Molecular Weight: 149.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 133
Flash Point (°C): 98
Specific Gravity (20/20): 1.16
Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Isocyanic Acid 4-Methoxyphenyl Ester
Formula:
C₈H₇NO₂ Boiling Pt: 133 °C Density: 1.16 (20 °C) Flash Pt: 98 °C |
MDL Number:
MFCD00002026 CAS Number: 5416-93-3 UN: 2206 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...