2-Methoxyphenyl isocyanate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Isocyanato-2-methoxybenzene

I0439-25G I0439-5G
TCI0439-25GEA 102 USD
TCI0439-25G TCI0439-5G
2-Methoxyphenyl isocyanate ≥98.0% (by GC)
2-Methoxyphenyl isocyanate

CAS Number: 700-87-8
MDL Number: MFCD00002004
Molecular Formula: C8H7NO2
Molecular Weight: 149.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 110
Specific Gravity (20/20): 1.16
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Isocyanic Acid 2-Methoxyphenyl Ester

Formula: C₈H₇NO₂
MW: 149.15 g/mol
Boiling Pt: 96 °C
Density: 1.16 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00002004
CAS Number: 700-87-8
UN: 2206
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab3adc86c8c3641dc4b01c60b620d79fa9702441&vid=ab3adc86c8c3641dc4b01c60b620d79fa9702441&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR