Benzyl isocyanate ≥99.0% (by GC)
Supplier: TCI America
CAS Number: 3173-56-6
MDL Number: MFCD00009701
Molecular Formula: C8H7NO
Molecular Weight: 133.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 45
Specific Gravity (20/20): 1.08
Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Isocyanic Acid Benzyl Ester
Formula:
C₈H₇NO Boiling Pt: 118 °C Density: 1.08 (20 °C) Flash Pt: 45 °C |
MDL Number:
MFCD00009701 CAS Number: 3173-56-6 UN: 3080 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...