p-Tolyl isothiocyanate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Methylphenyl isothiocyanate

I0393-25G I0393-5G
TCI0393-25GEA 168 USD
TCI0393-25G TCI0393-5G
p-Tolyl isothiocyanate ≥98.0% (by GC)
p-Tolyl isothiocyanate

CAS Number: 622-59-3
MDL Number: MFCD00004813
Molecular Formula: C8H7NS
Molecular Weight: 149.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 237
Flash Point (°C): 107
Freezing point (°C): 26


Other Product Names / Synonyms: Isothiocyanic Acid p-Tolyl Ester, 4-Methylphenyl Isothiocyanate

Formula: C₈H₇NS
MW: 149.22 g/mol
Boiling Pt: 237 °C
Flash Pt: 107 °C
MDL Number: MFCD00004813
CAS Number: 622-59-3
UN: 2923
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=261533ecfc597615fa4045dc9ab672f73e2f8a99&vid=261533ecfc597615fa4045dc9ab672f73e2f8a99&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR