1-Propyl isothiocyanate ≥98.0% (by GC)

Supplier: TCI America

I0388-25ML I0388-5ML
TCI0388-25MLEA 118.2 USD
TCI0388-25ML TCI0388-5ML
1-Propyl isothiocyanate ≥98.0% (by GC)
1-Propyl isothiocyanate

CAS Number: 628-30-8
MDL Number: MFCD00004823
Molecular Formula: C4H7NS
Molecular Weight: 101.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 153
Flash Point (°C): -15
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: Isothiocyanic Acid Propyl Ester

Formula: C₄H₇NS
Boiling Pt: 153 °C
Density: 0.97 (20 °C)
Flash Pt: –15 °C
MDL Number: MFCD00004823
CAS Number: 628-30-8
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f086065bb175113421c0f70c30cd818842ccee62&vid=f086065bb175113421c0f70c30cd818842ccee62&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR