4-Fluorophenyl isocyanate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4-Fluorophenylisocyanate, 1-Fluoro-4-isocyanatobenzene
I0339-25G
I0339-5G
TCI0339-25GEA
152
USD
TCI0339-25G
TCI0339-5G
4-Fluorophenyl isocyanate ≥98.0% (by GC)
4-Fluorophenyl isocyanate
CAS Number: 1195-45-5
MDL Number: MFCD00002023
Molecular Formula: C7H4FNO
Molecular Weight: 137.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 68
Flash Point (°C): 53
Specific Gravity (20/20): 1.22
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Isocyanic Acid 4-Fluorophenyl Ester
Formula:
C₇H₄FNO MW: 137.11 g/mol Boiling Pt: 68 °C Density: 1.22 (20 °C) Flash Pt: 53 °C |
MDL Number:
MFCD00002023 CAS Number: 1195-45-5 UN: 3080 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...