2-Fluorophenyl isocyanate ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 1-Fluoro-2-isocyanatobenzene

I0338-25G I0338-5G
TCI0338-25GEA 192.5 USD
TCI0338-25G TCI0338-5G
2-Fluorophenyl isocyanate ≥97.0% (by GC)
2-Fluorophenyl isocyanate

CAS Number: 16744-98-2
MDL Number: MFCD00001996
Molecular Formula: C7H4FNO
Molecular Weight: 137.11
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 65
Flash Point (°C): 50
Specific Gravity (20/20): 1.22
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Isocyanic Acid 2-Fluorophenyl Ester

Formula: C₇H₄FNO
MW: 137.11 g/mol
Boiling Pt: 65 °C
Density: 1.22 (20 °C)
Flash Pt: 50 °C
MDL Number: MFCD00001996
CAS Number: 16744-98-2
UN: 3080
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4d26511c2d2cde5300f83f46b34200f1d4e30ff&vid=b4d26511c2d2cde5300f83f46b34200f1d4e30ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR