2-Chloroethyl isocyanate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Isocyanic acid 2-Chloroethyl ester

I0293-25G I0293-5G
TCI0293-25GEA 105 USD
TCI0293-25G TCI0293-5G
2-Chloroethyl isocyanate ≥98.0% (by GC)
2-Chloroethyl isocyanate

CAS Number: 1943-83-5
MDL Number: MFCD00002043
Molecular Formula: C3H4ClNO
Molecular Weight: 105.52
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Flash Point (°C): 56
Specific Gravity (20/20): 1.23
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Isocyanic Acid 2-Chloroethyl Ester

Formula: C₃H₄ClNO
MW: 105.52 g/mol
Boiling Pt: 135 °C
Density: 1.23 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD00002043
CAS Number: 1943-83-5
UN: 3080
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e43dba3520556f5d4f2f667bc1b5ae02b80a637&vid=5e43dba3520556f5d4f2f667bc1b5ae02b80a637&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR