2-Propanesulfonyl chloride ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Isopropylsulphonyl chloride

I0291-25G I0291-5G
TCI0291-25GEA 165 USD
TCI0291-25G TCI0291-5G
2-Propanesulfonyl chloride ≥95.0% (by GC)
2-Propanesulphonyl chloride

CAS Number: 10147-37-2
MDL Number: MFCD00007453
Molecular Formula: C3H7ClO2S
Molecular Weight: 142.60
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 83
Flash Point (°C): 86
Specific Gravity (20/20): 1.28

Formula: C₃H₇ClO₂S
MW: 142.61 g/mol
Boiling Pt: 83 °C
Density: 1.28 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00007453
CAS Number: 10147-37-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7c2857a34be87135caacceb2560a44d58576b47&vid=c7c2857a34be87135caacceb2560a44d58576b47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR