Benzyl isothiocyanate ≥98.0%
Supplier: TCI America
Synonyms:
Benzylisothiocyanate
I0224-25G
I0224-5G
TCI0224-25GEA
137
USD
TCI0224-25G
TCI0224-5G
Benzyl isothiocyanate ≥98.0%
Benzyl isothiocyanate
CAS Number: 622-78-6
MDL Number: MFCD00004819
Molecular Formula: C8H7NS
Molecular Weight: 149.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 243
Flash Point (°C): 110
Specific Gravity (20/20): 1.13
Other Product Names / Synonyms: Isothiocyanic Acid Benzyl Ester
Formula:
C₈H₇NS Boiling Pt: 243 °C Density: 1.13 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00004819 CAS Number: 622-78-6 UN: 2206 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...