Diisopropyl sulfide ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Isopropyl sulphide
I0175-10ML
I0175-25ML
TCI0175-10MLEA
37
USD
TCI0175-10ML
TCI0175-25ML
Diisopropyl sulfide ≥98.0% (by GC)
Diisopropyl sulphide
CAS Number: 625-80-9
MDL Number: MFCD00008895
Molecular Formula: C6H14S
Molecular Weight: 118.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 121
Flash Point (°C): 7
Specific Gravity (20/20): 0.82
Other Product Names / Synonyms: Diisopropyl Sulfide
Formula:
C₆H₁₄S MW: 118.24 g/mol Boiling Pt: 121 °C Density: 0.82 (20 °C) Flash Pt: 7 °C |
MDL Number:
MFCD00008895 CAS Number: 625-80-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...