Isonicotinoyl chloride hydrochloride ≥95.0%
Supplier: TCI America
Synonyms:
Pyridine-4-carbonyl Chloride Hydrochloride, 4-Pyridinoyl chloride hydrochloride
I0144-250G
I0144-25G
TCI0144-250GEA
553
USD
TCI0144-250G
TCI0144-25G
Isonicotinoyl chloride hydrochloride ≥95.0%
Isonicotinoyl chloride hydrochloride
CAS Number: 39178-35-3
MDL Number: MFCD00012830
Molecular Formula: C6H4ClNO
Molecular Weight: 178.01
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 160
Other Product Names / Synonyms: Pyridine-4-carbonyl Chloride Hydrochloride
Formula:
C₆H₄ClNO Melting Pt: 160 °C |
MDL Number:
MFCD00012830 CAS Number: 39178-35-3 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...