Isohexane (mixture of isomeric branched chain Hexanes) ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Hexanes

I0134-25ML I0134-500ML
TCI0134-25MLEA 24 USD
TCI0134-25ML TCI0134-500ML
Isohexane (mixture of isomeric branched chain Hexanes) ≥98.0% (by GC)
Isohexane (mixture of isomeric branched chain Hexanes)

CAS Number: 73513-42-5
MDL Number: MFCD02179311
Molecular Formula: C6H14
Molecular Weight: 86.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): -30
Specific Gravity (20/20): 0.66

Formula: C₆H₁₄
Density: 0.66 (20 °C)
Flash Pt: –30 °C
MDL Number: MFCD02179311
CAS Number: 73513-42-5
ADR: 3,II

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR