4-(Indol-3-yl)butyric acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 133-32-4
MDL Number: MFCD00005664
Molecular Formula: C12H13NO2
Molecular Weight: 203.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 126
Product Categories: Bioscience, Auxins
Other Product Names / Synonyms: IBA
Formula:
C₁₂H₁₃NO₂ Melting Pt: 126 °C |
MDL Number:
MFCD00005664 CAS Number: 133-32-4 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...