2,2,3,3,4,4,4-Heptafluorobutylamine ≥95.0% (by titrimetric analysis)

Supplier: TCI America

H1300-1G H1300-5G
TCH1300-1GEA 52 USD
TCH1300-1G TCH1300-5G
2,2,3,3,4,4,4-Heptafluorobutylamine ≥95.0% (by titrimetric analysis)
2,2,3,3,4,4,4-Heptafluorobutylamine

CAS Number: 374-99-2
MDL Number: MFCD00014817
Molecular Formula: C4H4F7N
Molecular Weight: 199.07
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Boiling point (°C): 69
Specific Gravity (20/20): 1.50


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1H,1H-Perfluorobutylamine

Formula: C₄H₄F₇N
MW: 199.07 g/mol
Boiling Pt: 69 °C
Density: 1.50 (20 °C)
MDL Number: MFCD00014817
CAS Number: 374-99-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60c09a46cb672697289ca787065278406b0cf0e7&vid=60c09a46cb672697289ca787065278406b0cf0e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR