1,1,1,3,5,5,5-Heptamethyltrisiloxane ≥98.0%

Supplier: TCI America

Synonyms: Methylbis(trimethylsilyloxy)silane

H1267-250ML H1267-25ML
TCH1267-250MLEA 115 USD
TCH1267-250ML TCH1267-25ML
1,1,1,3,5,5,5-Heptamethyltrisiloxane ≥98.0%
1,1,1,3,5,5,5-Heptamethyltrisiloxane

CAS Number: 1873-88-7
MDL Number: MFCD00048000
Molecular Formula: C7H22O2Si3
Molecular Weight: 222.51
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 22
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Methylbis(trimethylsilyloxy)silane

Formula: C₇H₂₂O₂Si₃
MW: 222.51
Boiling Pt: 142 °C
Density: 0.82 (20 °C)
Flash Pt: 22 °C
MDL Number: MFCD00048000
CAS Number: 1873-88-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e214975f2b7a2adb3239a3fc10b1df309dfb9fe&vid=1e214975f2b7a2adb3239a3fc10b1df309dfb9fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR