5-Hexynoic acid ≥96.0% (by GC, titration analysis)

Supplier: TCI America

H0882-25G H0882-5G
TCH0882-25GEA 403 USD
TCH0882-25G TCH0882-5G
5-Hexynoic acid ≥96.0% (by GC, titration analysis)
5-Hexynoic acid

CAS Number: 53293-00-8
MDL Number: MFCD00066346
Molecular Formula: C6H8O2
Molecular Weight: 112.13
Purity/Analysis Method: >96.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 124
Flash Point (°C): 110
Specific Gravity (20/20): 1.03

Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 124 °C
Density: 1.03 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00066346
CAS Number: 53293-00-8
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fedb1284951b43397b5539bd448dc62a7dcdb644&vid=fedb1284951b43397b5539bd448dc62a7dcdb644&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR