5-Hexenoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
5-Hexenoicacid
H0875-25G
H0875-5G
TCH0875-25GEA
536
USD
TCH0875-25G
TCH0875-5G
5-Hexenoic acid ≥98.0% (by GC, titration analysis)
5-Hexenoic acid
CAS Number: 1577-22-6
MDL Number: MFCD00046558
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 203
Flash Point (°C): 104
Specific Gravity (20/20): 0.96
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 203 °C Density: 0.96 (20 °C) Flash Pt: 104 °C |
MDL Number:
MFCD00046558 CAS Number: 1577-22-6 UN: 3265 ADR: 8,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...