4,4'-(Hexafluoroisopropylidene)diphthalic anhydride ≥98.0%
Supplier: TCI America
Synonyms:
5,5'-(Perfluoropropane-2,2-diyl)bis(isobenzofuran-1,3-dione), 2,2-Bis(1,3-dihydro-1,3-dioxobenzo[c]furan-5-yl)hexafluoropropane
H0771-25G
H0771-5G
H0771-100G
TCH0771-25GEA
145
USD
TCH0771-25G
TCH0771-5G
TCH0771-100G
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride ≥98.0%
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride
CAS Number: 1107-00-2
MDL Number: MFCD00039143
Molecular Formula: C19H6F6O6
Molecular Weight: 444.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 246
Formula:
C₁₉H₆F₆O₆ Melting Pt: 246 °C |
MDL Number:
MFCD00039143 CAS Number: 1107-00-2 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...