2,4-Hexadiyne-1,6-diol ≥99.0% (by GC)
Supplier: TCI America
CAS Number: 3031-68-3
MDL Number: MFCD00020619
Molecular Formula: C6H6O2
Molecular Weight: 110.11
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 113
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Langmuir-Blodgett(LB) Film Forming Agents
Other Product Names / Synonyms: 1,6-Dihydroxy-2,4-hexadiyne
Formula:
C₆H₆O₂ Melting Pt: 113 °C |
MDL Number:
MFCD00020619 CAS Number: 3031-68-3 UN: 1325 ADR: 4.1,III |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...