trans-2-Heptenal ≥95.0% (by GC)

Supplier: TCI America

H0579-25ML H0579-5ML
TCH0579-25MLEA 203 USD
TCH0579-25ML TCH0579-5ML
trans-2-Heptenal ≥95.0% (by GC)
trans-2-Heptenal

CAS Number: 18829-55-5
MDL Number: MFCD00007010
Molecular Formula: C7H12O
Molecular Weight: 112.17
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 91
Flash Point (°C): 54
Specific Gravity (20/20): 0.85

Formula: C₇H₁₂O
Boiling Pt: 91 °C
Density: 0.85 (20 °C)
Flash Pt: 54 °C
MDL Number: MFCD00007010
CAS Number: 18829-55-5
UN: 1988
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=134f0d7f3709e1cf1dc6c342ea793d630d5c0a78&vid=4863443a9f7e0592ecc66baa58a900c772499698&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR