3,4-Hexanedione ≥96.0% (by GC)
Supplier: TCI America
CAS Number: 4437-51-8
MDL Number: MFCD00010237
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 27
Freezing point (°C): -10
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: Dipropionyl
Formula:
C₆H₁₀O₂ Boiling Pt: 130 °C Density: 0.95 (20 °C) Flash Pt: 27 °C |
MDL Number:
MFCD00010237 CAS Number: 4437-51-8 UN: 1224 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...