2,4-Hexadiyne ≥98.0% (by GC)

Supplier: TCI America

H0502-1G H0502-5G
TCH0502-1GEA 136 USD
TCH0502-1G TCH0502-5G
2,4-Hexadiyne ≥98.0% (by GC)
2,4-Hexadiyne

CAS Number: 2809-69-0
MDL Number: MFCD00041617
Molecular Formula: C6H6
Molecular Weight: 78.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 129
Melting point (°C): 66
Flash Point (°C): 35

Formula: C₆H₆
Boiling Pt: 129 °C
Melting Pt: 66 °C
Flash Pt: 35 °C
MDL Number: MFCD00041617
CAS Number: 2809-69-0
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ad11d84e512e7c7f94f375922db6070ad5efaf0&vid=8ad11d84e512e7c7f94f375922db6070ad5efaf0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR