3-Hydroxy-3-methylbutanone ≥95.0%

Supplier: TCI America

Synonyms: 3-Hydroxy-3-methyl-2-butanone, 3-Hydroxy-3-methylbutan-2-one

H0471-25ML H0471-100ML
TCH0471-25MLEA 120.3 USD
TCH0471-25ML TCH0471-100ML
3-Hydroxy-3-methylbutanone ≥95.0%
3-Hydroxy-3-methylbutanone

CAS Number: 115-22-0
MDL Number: MFCD00004460
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 138
Flash Point (°C): 42
Specific Gravity (20/20): 0.96
Storage Temperature: 0-10°C

Formula: C₅H₁₀O₂
Boiling Pt: 138 °C
Density: 0.96 (20 °C)
Flash Pt: 42 °C
MDL Number: MFCD00004460
CAS Number: 115-22-0
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a6c95637c8e688c9946b73355097a844c6177d2&vid=1a6c95637c8e688c9946b73355097a844c6177d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR