trans-2-Hexenoic acid ≥98.0% (by GC)

Supplier: TCI America

Synonyms: trans-Hex-2-enoic acid

H0383-250G H0383-25G
TCH0383-250GEA 301 USD
TCH0383-250G TCH0383-25G
trans-2-Hexenoic acid ≥98.0% (by GC)
trans-2-Hexenoic acid

CAS Number: 13419-69-7
MDL Number: MFCD00002705
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 118
Flash Point (°C): 125
Freezing point (°C): 32

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 118 °C
Flash Pt: 125 °C
MDL Number: MFCD00002705
CAS Number: 13419-69-7
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48f633555afd462afe595c8b3e28903dc14ab4f3&vid=48f633555afd462afe595c8b3e28903dc14ab4f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR