2-Hydrazinobenzothiazole ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Hydrazinylbenzo[d]thiazole

H0177-25G H0177-5G
TCH0177-25GEA 143 USD
TCH0177-25G TCH0177-5G
2-Hydrazinobenzothiazole ≥99.0% (by titrimetric analysis)
2-Hydrazinobenzothiazole

CAS Number: 615-21-4
MDL Number: MFCD00041849
Molecular Formula: C7H7N3S
Molecular Weight: 165.21
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 202


Other Product Names / Synonyms: 2-Benzothiazolylhydrazine

Formula: C₇H₇N₃S
MW: 165.21 g/mol
Melting Pt: 202 °C
MDL Number: MFCD00041849
CAS Number: 615-21-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8dccaf8412922a5c3b9c786d178b196360ca1c53&vid=8dccaf8412922a5c3b9c786d178b196360ca1c53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR