n-Hexylbenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Phenylhexane, Hexylbenzene

H0136-250ML H0136-25ML
TCH0136-250MLEA 556 USD
TCH0136-250ML TCH0136-25ML
n-Hexylbenzene ≥98.0% (by GC)
n-Hexylbenzene

CAS Number: 1077-16-3
MDL Number: MFCD00009526
Molecular Formula: C12H18
Molecular Weight: 162.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 226
Melting point (°C): -61
Flash Point (°C): 83
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: 1-Phenylhexane

Formula: C₁₂H₁₈
MW: 162.27 g/mol
Boiling Pt: 226 °C
Melting Pt: -61 °C
Density: 0.86 (20 °C)
Flash Pt: 83 °C
MDL Number: MFCD00009526
CAS Number: 1077-16-3
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=650eb9f170f46525b9ec4a55eb7cb293197a1aff&vid=ec09958d4d0a9613a8caf086f9d6d00f2d45f165&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR