3-Hexanone ≥98.0% (by GC)
Supplier: TCI America
H0115-25ML
H0115-5ML
TCH0115-25MLEA
111
USD
TCH0115-25ML
TCH0115-5ML
3-Hexanone ≥98.0% (by GC)
3-Hexanone
CAS Number: 589-38-8
MDL Number: MFCD00009402
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 124
Flash Point (°C): 14
Specific Gravity (20/20): 0.82
Other Product Names / Synonyms: Ethyl Propyl Ketone
Formula:
C₆H₁₂O MW: 100.16 g/mol Boiling Pt: 124 °C Density: 0.82 (20 °C) Flash Pt: 14 °C |
MDL Number:
MFCD00009402 CAS Number: 589-38-8 UN: 1224 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...