Heptylbenzene ≥97.0% (by GC)

Supplier: TCI America

Synonyms: N-heptylbenzene, 1-Phenylheptane

H0046-25ML H0046-500ML
TCH0046-25MLEA 98.72 USD
TCH0046-25ML TCH0046-500ML
Heptylbenzene ≥97.0% (by GC)
Heptylbenzene

CAS Number: 1078-71-3
MDL Number: MFCD00009545
Molecular Formula: C13H20
Molecular Weight: 176.30
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 235
Flash Point (°C): 95
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: 1-Phenylheptane

Formula: C₁₃H₂₀
MW: 176.3 g/mol
Boiling Pt: 235 °C
Density: 0.86 (20 °C)
Flash Pt: 95 °C
MDL Number: MFCD00009545
CAS Number: 1078-71-3
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8edf4d071576b2e90cf9e61b304b29da5de0edd2&vid=76186fe99abf320c5583952ecc01c0e9c69504e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR