Heptafluorobutyric acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Perfluorobutyric acid, HFBA, Heptafluorobutanoic acid

H0024-100G H0024-25G
TCH0024-100GEA 330 USD
TCH0024-100G TCH0024-25G
Heptafluorobutyric acid ≥98.0% (by GC, titration analysis)
Heptafluorobutyric acid

CAS Number: 375-22-4
MDL Number: MFCD00004171
Molecular Formula: C4HF7O2
Molecular Weight: 214.04
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 120
Melting point (°C): -20
Specific Gravity (20/20): 1.65


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Analytical Science, Acylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: Perfluorobutyric Acid

Formula: C₄HF₇O₂
MW: 214.04 g/mol
Boiling Pt: 120 °C
Melting Pt: –20 °C
Density: 1.65 (20 °C)
MDL Number: MFCD00004171
CAS Number: 375-22-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8336b146ac222f7aa429e0b250ed8fb623a9e298&vid=8336b146ac222f7aa429e0b250ed8fb623a9e298&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR