(S)-(-)-2,3-Epoxy-1-propanol ≥96.0% (by GC)

Supplier: TCI America

Synonyms: (S)-(-)-Glycidol, (S)-Oxiran-2-ylmethanol

G0364-25G G0364-5G
TCG0364-25GEA 250 USD
TCG0364-25G TCG0364-5G
(S)-(-)-2,3-Epoxy-1-propanol ≥96.0% (by GC)
(S)-(-)-2,3-Epoxy-1-propanol

CAS Number: 60456-23-7
MDL Number: MFCD00074874
Molecular Formula: C3H6O2
Molecular Weight: 74.08
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 67
Flash Point (°C): 72
Specific Gravity (20/20): 1.12
Specific rotation [a]20/D: -15 deg (neat)
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-1-Hydroxy-2,3-epoxypropane, (S)-(-)-2-(Hydroxymethyl)oxirane

Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 67 °C
Density: 1.12 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00074874
CAS Number: 60456-23-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05d4130528c17f1c2359dc6ec270aca8985a2385&vid=05d4130528c17f1c2359dc6ec270aca8985a2385&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR