(R)-(+)-2,3-Epoxy-1-propanol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (R)-(+)-2-(Hydroxymethyl)oxirane, (R)-(+)-1-Hydroxy-2,3-epoxypropane, (R)-Oxiran-2-ylmethanol, (R)-(+)-Glycidol

G0363-25G G0363-5G
TCG0363-25GEA 280.56 USD
TCG0363-25G TCG0363-5G
(R)-(+)-2,3-Epoxy-1-propanol ≥98.0% (by GC)
(R)-(+)-2,3-Epoxy-1-propanol

CAS Number: 57044-25-4
MDL Number: MFCD00074873
Molecular Formula: C3H6O2
Molecular Weight: 74.08
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 57
Flash Point (°C): 72
Specific Gravity (20/20): 1.12
Specific rotation [a]20/D: 15 deg (neat)
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(+)-1-Hydroxy-2,3-epoxypropane, (R)-(+)-2-(Hydroxymethyl)oxirane

Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 57 °C
Density: 1.12 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00074873
CAS Number: 57044-25-4
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be2bb5e918068ad6e343b995cbc090c7b5669e85&vid=be2bb5e918068ad6e343b995cbc090c7b5669e85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR