(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulfonate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (S)-(+)-Glycidyl nosylate, (S)-Oxiran-2-ylmethyl 3-nitrobenzenesulphonate, Glycidyl (S)-(+)-3-nitrobenzenesulphonate

G0287-25G G0287-5G
TCG0287-25GEA 230.2 USD
TCG0287-25G TCG0287-5G
(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulfonate ≥98.0% (by GC)
(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulfonate

CAS Number: 115314-14-2
MDL Number: MFCD00064582
Molecular Formula: C9H9NO6S
Molecular Weight: 259.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 65
Flash Point (°C): 150
Specific rotation [a]20/D: 23 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: 3-Nitrobenzenesulfonic Acid (S)-Glycidyl Ester

Formula: C9H9NO6S
MW: 259.23 g/mol
Melting Pt: 65 °C
Flash Pt: 150 °C
MDL Number: MFCD00064582
CAS Number: 115314-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3743cb44b9cbf2f2c0a3d503d11447ab1a6ed9e&vid=e3743cb44b9cbf2f2c0a3d503d11447ab1a6ed9e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR