(R)-Glycidyl-3-nitrobenzenesulfonate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(R)-(-)-Glycidyl nosylate, 3-Nitrobenzenesulphonic acid (R)-Glycidyl Ester, (R)-Oxiran-2-ylmethyl 3-nitrobenzenesulphonate
G0286-25G
G0286-5G
TCG0286-25GEA
179
USD
TCG0286-25G
TCG0286-5G
(R)-Glycidyl-3-nitrobenzenesulfonate ≥98.0% (by GC)
(R)-Glycidyl-3-nitrobenzenesulfonate
CAS Number: 115314-17-5
MDL Number: MFCD00013436
Molecular Formula: C9H9NO6S
Molecular Weight: 259.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 63
Flash Point (°C): 150
Specific rotation [a]20/D: -22.5 deg (C=1, CHCl3)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: 3-Nitrobenzenesulfonic Acid (R)-Glycidyl Ester
Formula:
C₉H₉NO₆S MW: 259.23 g/mol Melting Pt: 63 °C Flash Pt: 150 °C |
MDL Number:
MFCD00013436 CAS Number: 115314-17-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...