(S)-(+)-Glycidyl methyl ether ≥97.0% (by GC)

Supplier: TCI America

G0281-25G G0281-5G
TCG0281-25GEA 466.6 USD
TCG0281-25G TCG0281-5G
(S)-(+)-Glycidyl methyl ether ≥97.0% (by GC)
(S)-(+)-Glycidyl methyl ether

CAS Number: 64491-68-5
MDL Number: MFCD00273366
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 17
Specific Gravity (20/20): 0.98
Specific rotation [a]20/D: 14 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-1-Methoxy-2,3-epoxypropane, (S)-2-(Methoxymethyl)oxirane

Formula: C₄H₈O₂
Density: 0.98 (20 °C)
Flash Pt: 17 °C
MDL Number: MFCD00273366
CAS Number: 64491-68-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ca4f3b629ee8812fdc0c035d209704f80b01ca7&vid=5ca4f3b629ee8812fdc0c035d209704f80b01ca7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR