(R)-(-)-Glycidyl methyl ether ≥97.0% (by GC)
Supplier: TCI America
G0280-25G
G0280-5G
TCG0280-25GEA
559
USD
TCG0280-25G
TCG0280-5G
(R)-(-)-Glycidyl methyl ether ≥97.0% (by GC)
(R)-(-)-Glycidyl methyl ether
CAS Number: 64491-70-9
MDL Number: MFCD00273372
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 20
Specific Gravity (20/20): 0.99
Specific rotation [a]20/D: -14 deg (neat)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-1-Methoxy-2,3-epoxypropane, (R)-2-(Methoxymethyl)oxirane
Formula:
C₄H₈O₂ Density: 0.99 (20 °C) Flash Pt: 20 °C |
MDL Number:
MFCD00273372 CAS Number: 64491-70-9 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...