2-Fluoro-5-(trifluoromethyl)phenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0780-25G F0780-5G
TCF0780-25GEA 241.7 USD
TCF0780-25G TCF0780-5G
2-Fluoro-5-(trifluoromethyl)phenol ≥98.0% (by GC, titration analysis)
2-Fluoro-5-(trifluoromethyl)phenol

CAS Number: 141483-15-0
MDL Number: MFCD00061286
Molecular Formula: C7H4F4O
Molecular Weight: 180.10
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 146
Flash Point (°C): 57
Specific Gravity (20/20): 1.45


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-3-hydroxybenzotrifluoride

Formula: C₇H₄F₄O
Boiling Pt: 146 °C
Density: 1.45 (20 °C)
Flash Pt: 57 °C
MDL Number: MFCD00061286
CAS Number: 141483-15-0
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06d8ef9863209f0e7e0dbad2a6f0189b30a634f2&vid=06d8ef9863209f0e7e0dbad2a6f0189b30a634f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR