4-Fluoro-3-(trifluoromethyl)phenol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 61721-07-1
MDL Number: MFCD00043875
Molecular Formula: C7H4F4O
Molecular Weight: 180.10
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 86
Flash Point (°C): 79
Freezing point (°C): 17
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoro-5-hydroxybenzotrifluoride, alpha,alpha,alpha,4-Tetrafluoro-m-cresol
Formula:
C₇H₄F₄O Boiling Pt: 86 °C Flash Pt: 79 °C |
MDL Number:
MFCD00043875 CAS Number: 61721-07-1 UN: 3265 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...