Fluoxetine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-Methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propylamine Hydrochloride

F0750-1G F0750-5G
TCF0750-1GEA 115.07 USD
TCF0750-1G TCF0750-5G
Fluoxetine hydrochloride ≥98.0% (by HPLC, titration analysis)
Fluoxetine hydrochloride

CAS Number: 56296-78-7
MDL Number: MFCD00214288
Molecular Formula: C17H18F3NO
Molecular Weight: 345.79
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 159


Product Categories: Bioscience, Regenerative Medicine Research Reagents, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: N-Methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propylamine Hydrochloride

Formula: C₁₇H₁₈F₃NO
MW: 345.79 g/mol
Melting Pt: 159 °C
MDL Number: MFCD00214288
CAS Number: 56296-78-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c87e4b1f92f62258dad72866b938f3465c943bef&vid=c87e4b1f92f62258dad72866b938f3465c943bef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR