2-Fluoro-6-(trifluoromethyl)benzonitrile ≥98.0% (by GC)

Supplier: TCI America

F0701-1G F0701-5G
TCF0701-1GEA 39 USD
TCF0701-1G TCF0701-5G
2-Fluoro-6-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
2-Fluoro-6-(trifluoromethyl)benzonitrile

CAS Number: 133116-83-3
MDL Number: MFCD00061153
Molecular Formula: C8H3F4N
Molecular Weight: 189.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 28
Flash Point (°C): 99


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Cyano-1-fluoro-3-(trifluoromethyl)benzene

Formula: C₈H₃F₄N
Melting Pt: 28 °C
Flash Pt: 99 °C
MDL Number: MFCD00061153
CAS Number: 133116-83-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b5c3e630e439e36eec921eb2bfd188756648481&vid=3b5c3e630e439e36eec921eb2bfd188756648481&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR