3-Fluoro-4-methoxybenzonitrile ≥98.0% (by GC)

Supplier: TCI America

F0671-1G F0671-5G
TCF0671-1GEA 34.5 USD
TCF0671-1G TCF0671-5G
3-Fluoro-4-methoxybenzonitrile ≥98.0% (by GC)
3-Fluoro-4-methoxybenzonitrile

CAS Number: 331-62-4
MDL Number: MFCD00215835
Molecular Formula: C8H6FNO
Molecular Weight: 151.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 98
Melting point (°C): 101


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Cyano-2-fluoroanisole

Formula: C₈H₆FNO
MW: 151.14 g/mol
Boiling Pt: 98 °C
Melting Pt: 101 °C
MDL Number: MFCD00215835
CAS Number: 331-62-4
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fdc6ca0073ec54275e8c4e8db11a90de5b7be19&vid=6fdc6ca0073ec54275e8c4e8db11a90de5b7be19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR