2-Fluoro-5-methoxybenzonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Cyan-2-fluor-5-methoxybenzene

F0642-1G F0642-5G
TCF0642-1GEA 29 USD
TCF0642-1G TCF0642-5G
2-Fluoro-5-methoxybenzonitrile ≥98.0% (by GC)
2-Fluoro-5-methoxybenzonitrile

CAS Number: 127667-01-0
MDL Number: MFCD00142921
Molecular Formula: C8H6FNO
Molecular Weight: 151.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 82


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Cyano-4-fluoroanisole

Formula: C₈H₆FNO
MW: 151.14 g/mol
Melting Pt: 82 °C
MDL Number: MFCD00142921
CAS Number: 127667-01-0
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d428d9910832353390cdccd1622c53910b0e175d&vid=d428d9910832353390cdccd1622c53910b0e175d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR