4,5-Dimethoxy-2-fluorobenzonitrile ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-Fluoro-4,5-dimethoxybenzonitrile
F0641-1G
F0641-5G
TCF0641-1GEA
79.5
USD
TCF0641-1G
TCF0641-5G
4,5-Dimethoxy-2-fluorobenzonitrile ≥98.0% (by GC)
4,5-Dimethoxy-2-fluorobenzonitrile
CAS Number: 119396-88-2
MDL Number: MFCD00143236
Molecular Formula: C9H8FNO2
Molecular Weight: 181.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 143
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Cyano-5-fluoroveratrole
Formula:
C₉H₈FNO₂ MW: 181.1637 g/mol Melting Pt: 143 °C |
MDL Number:
MFCD00143236 CAS Number: 119396-88-2 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...